missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Paraneoplastic antigen-like protein 1
A DNA sequence encoding the Homo sapiens (Human) Paraneoplastic antigen-like protein 5, was expressed in the hosts and tags indicated.
Marke: enQuireBio™ QP7986-ec-200ug
Weitere Details : Weight : 0.01000kg
Spezifikation
NP_001096620.1 | |
Paraneoplastic antigen-like protein 1 | |
Research Use Only | |
PNMA5 | |
Recombinant Protein | |
E. coli | |
TLLEDWCKGM DMDPRKALLI VGIPMECSEV EIQDTVKAGL QPLCAYRVLG RMFRREDNAK AVFIELADTV NYTTLPSHIP GKGGSWEVVV KPRNPDDEFL SRLNYFLKDE GRSMTDVARA LGCCSLPAES LDAEVMPQVR SPPLEPPKES MWYRKLKVFS GTASPSPGEE TFEDWLEQVT EIMPIWQVSE VEKRRRLLES LRGPALSIMR VLQANNDSIT VEQCLDALKQ IFGDKEDFRA SQFRFLQTSP KIGEKVSTFL LRLEPLLQKA VHKSPLSVRS TDMIRLKHLL ARVAMTPALR GKLELLDQRG CPPNFLELMK LIRDEEEWEN TEAVMKNKEK PSGRGRGASG RQARAEASVS APQATVQARS FSDSSPQTIQ GGLPPLVKRR RLLGSESTRG EDHGQATYPK AENQTPGREG PQAAGEELGN EAGAGAMSH | |
Greater than 90% as determined by SDS-PAGE. |
114824 | |
200 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |