missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human PCK1 Protein
A DNA sequence encoding the Homo sapiens (Human) PCK1, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6469-ec-500ug
Additional Details : Weight : 0.01000kg
Specifications
NP_002582.3 | |
PCK1 Protein | |
Research Use Only | |
PCK1 | |
Recombinant Protein | |
E. coli | |
MPPQLQNGLN LSAKVVQGSL DSLPQAVREF LENNAELCQP DHIHICDGSE EENGRLLGQM EEEGILRRLK KYDNCWLALT DPRDVARIES KTVIVTQEQR DTVPIPKTGL SQLGRWMSEE DFEKAFNARF PGCMKGRTMY VIPFSMGPLG SPLSKIGIEL TDSPYVVASM RIMTRMGTPV LEAVGDGEFV KCLHSVGCPL PLQKPLVNNW PCNPELTLIA HLPDRREIIS FGSGYGGNSL LGKKCFALRM ASRLAKEEGW LAEHMLILGI TNPEGEKKYL AAAFPSACGK TNLAMMNPSL PGWKVECVGD DIAWMKFDAQ GHLRAINPEN GFFGVAPGTS VKTNPNAIKT IQKNTIFTNV AETSDGGVYW EGIDEPLASG VTITSWKNKE WSSEDGEPCA HPNSRFCTPA SQCPIIDAAW ESPEGVPIEG IIFGGRRPAG VPLVYEALSW QHGVFVGAAM RSEATAAAEH KGKIIMHDPF AMRPFFGYNF GKYLAHWLSM AQHPAAKLPK IFHVNWFRKD KEGKFLWPGF GENSRVLEWM FNRIDGKAST KLTPIGYIPK EDALNLKGLG HINMMELFSI SKEFWEKEVE DIEKYLEDQV NADLPCEIER EILALKQRIS QM | |
Greater than 90% as determined by SDS-PAGE. |
5105 | |
500 μg | |
Not determined. | |
Full Length | |
Human | |
His | |
Tris-based buffer, 50% glycerol |