missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human PDZK1 Protein
A DNA sequence encoding the Homo sapiens (Human) PDZK1, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP7705-ec-500ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001188254.1 | |
PDZK1 Protein | |
500 μg | |
Not determined. | |
Full Length | |
Human | |
His | |
Tris-based buffer, 50% glycerol |
5174 | |
Greater than 90% as determined by SDS-PAGE. | |
Research Use Only | |
PDZK1 | |
Recombinant Protein | |
E. coli | |
MTSTFNPREC KLSKQEGQNY GFFLRIEKDT EGHLVRVVEK CSPAEKAGLQ DGDRVLRING VFVDKEEHMQ VVDLVRKSGN SVTLLVLDGD SYEKAVKTRV DLKELGQSQK EQGLSDNILS PVMNGGVQTW TQPRLCYLVK EGGSYGFSLK TVQGKKGVYM TDITPQGVAM RAGVLADDHL IEVNGENVED ASHEEVVEKV KKSGSRVMFL LVDKETDKRH VEQKIQFKRE TASLKLLPHQ PRIVEMKKGS NGYGFYLRAG SEQKGQIIKD IDSGSPAEEA GLKNNDLVVA VNGESVETLD HDSVVEMIRK GGDQTSLLVV DKETDNMYRL AHFSPFLYYQ SQELPNGSVK EAPAPTPTSL EVSSPPDTTE EVDHKPKLCR LAKGENGYGF HLNAIRGLPG SFIKEVQKGG PADLAGLEDE DVIIEVNGVN VLDEPYEKVV DRIQSSGKNV TLLVCGKKAY DYFQAKKIPI VSSLADPLDT PPDSKEGIVV ESNHDSHMAK ERAHSTASHS SSNSEDTEM |