missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Peptidyl-tRNA hydrolase ICT1, mitochondrial Protein
A DNA sequence encoding the Homo sapiens (Human) Peptidyl-tRNA hydrolase ICT1, mitochondrial, was expressed in the hosts and tags indicated.
1640.00 SEK - 26990.00 SEK
Specifications
Accession Number | NP_001290194.1 |
---|---|
Gene ID (Entrez) | 3396 |
Name | Peptidyl-tRNA hydrolase ICT1, mitochondrial Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16059200
|
enQuireBio™
QP7530-EC-100UG |
100 μg |
5980.00 SEK
100µg |
Estimated Shipment: 30-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16079200
|
enQuireBio™
QP7530-EC-1MG |
1 mg |
26990.00 SEK
1mg |
Estimated Shipment: 30-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16069200
|
enQuireBio™
QP7530-EC-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16089200
|
enQuireBio™
QP7530-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16099200
|
enQuireBio™
QP7530-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16009210
|
enQuireBio™
QP7530-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_001290194.1 | |
Peptidyl-tRNA hydrolase ICT1, mitochondrial Protein | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
3396 | |
Research Use Only | |
ICT1 | |
Recombinant Protein | |
E. coli | |
LHKQKDGTEF KSIYSLDKLY PESQGSDTAW RVPNGAKQAD SDIPLDRLTI SYCRSSGPGG QNVNKVNSKA EVRFHLATAE WIAEPVRQKI AITHKNKINR LGELILTSES SRYQFRNLAD CLQKIRDMIT EASQTPKEPT KEDVKLHRIR IENMNRERLR QKRIHSAVKT SRRVDMD | |
Greater than 90% as determined by SDS-PAGE. |