missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Peroxisomal biogenesis factor 19 Protein
A DNA sequence encoding the Homo sapiens (Human) Peroxisomal biogenesis factor 19, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6483-ec-10ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001180573.1 | |
Peroxisomal biogenesis factor 19 Protein | |
Research Use Only | |
PEX19 | |
Recombinant Protein | |
E. coli | |
AAAEEGCSVG AEADRELEEL LESALDDFDK AKPSPAPPST TTAPDASGPQ KRSPGDTAKD ALFASQEKFF QELFDSELAS QATAEFEKAM KELAEEEPHL VEQFQKLSEA AGRVGSDMTS QQEFTSCLKE TLSGLAKNAT DLQNSSMSEE ELTKAMEGLG MDEGDGEGNI LPIMQSIMQN LLSKDVLYPS LKEITEKYPE WLQSHRESLP PEQFEKYQEQ HSVMCKICEQ FEAETPTDSE TTQKARFEMV LDLMQQLQDL GHPPKELAGE MPPGLNFDLD ALNLSGPPGA SGEQC | |
Greater than 80% as determined by SDS-PAGE. |
5824 | |
10 μg | |
Not determined. | |
Full Length | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |