missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Phosphoglucomutase-1 Protein
A DNA sequence encoding the Homo sapiens (Human) Phosphoglucomutase-1, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6486-ec-200ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001166289.1 | |
Phosphoglucomutase-1 Protein | |
Research Use Only | |
PGM1 | |
Recombinant Protein | |
E. coli | |
MVKIVTVKTQ AYQDQKPGTS GLRKRVKVFQ SSANYAENFI QSIISTVEPA QRQEATLVVG GDGRFYMKEA IQLIARIAAA NGIGRLVIGQ NGILSTPAVS CIIRKIKAIG GIILTASHNP GGPNGDFGIK FNISNGGPAP EAITDKIFQI SKTIEEYAVC PDLKVDLGVL GKQQFDLENK FKPFTVEIVD SVEAYATMLR SIFDFSALKE LLSGPNRLKI RIDAMHGVVG PYVKKILCEE LGAPANSAVN CVPLEDFGGH HPDPNLTYAA DLVETMKSGE HDFGAAFDGD GDRNMILGKH GFFVNPSDSV AVIAANIFSI PYFQQTGVRG FARSMPTSGA LDRVASATKI ALYETPTGWK FFGNLMDASK LSLCGEESFG TGSDHIREKD GLWAVLAWLS ILATRKQSVE DILKDHWQKY GRNFFTRYDY EEVEAEGANK MMKDLEALMF DRSFVGKQFS ANDKVYTVEK ADNFEYSDPV DGSISRNQGL RLIFTDGSRI VFRLSGTGSA GATIRLYIDS YEKDVAKINQ DPQVMLAPLI SIALKVSQLQ ERTGRTAPTV IT | |
Greater than 90% as determined by SDS-PAGE. |
5236 | |
200 μg | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |