missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Phospholipid scramblase 1 Protein

A DNA sequence encoding the Homo sapiens (Human) Phospholipid scramblase 1, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP6506-ec-50ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16073780

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Description

Description

Specifications

Specifications

NP_066928.1
Phospholipid scramblase 1 Protein
Research Use Only
PLSCR1
Recombinant Protein
E. coli
ESTGSQEQKS GVW
Greater than 80% as determined by SDS-PAGE.
5359
50 μg
Not determined.
Full Length
Human
GST
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers