missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Phytanoyl-CoA dioxygenase, peroxisomal Protein
A DNA sequence encoding the Homo sapiens (Human) Phytanoyl-CoA dioxygenase, peroxisomal, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6492-ec-50ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001032626.1 | |
Phytanoyl-CoA dioxygenase, peroxisomal Protein | |
Research Use Only | |
PHYH | |
Recombinant Protein | |
E. coli | |
SGTISSASFH PQQFQYTLDN NVLTLEQRKF YEENGFLVIK NLVPDADIQR FRNEFEKICR KEVKPLGLTV MRDVTISKSE YAPSEKMITK VQDFQEDKEL FRYCTLPEIL KYVECFTGPN IMAMHTMLIN KPPDSGKKTS RHPLHQDLHY FPFRPSDLIV CAWTAMEHIS RNNGCLVVLP GTHKGSLKPH DYPKWEGGVN KMFHGIQDYE ENKARVHLVM EKGDTVFFHP LLIHGSGQNK TQGFRKAISC HFASADCHYI DVKGTSQENI EKEVVGIAHK FFGAENSVNL KDIWMFRARL VKGERTNL | |
Greater than 80% as determined by SDS-PAGE. |
5264 | |
50 μg | |
Not determined. | |
Full Length | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |