missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human PIK3R5 Protein
A DNA sequence encoding the Homo sapiens (Human) PIK3R5, was expressed in the hosts and tags indicated.
1580.00 SEK - 26990.00 SEK
Specifications
Accession Number | NP_001136105.1 |
---|---|
Gene ID (Entrez) | 23533 |
Name | PIK3R5 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16051531
|
enQuireBio™
QP7909-EC-100UG |
100 μg |
5980.00 SEK
100µg |
Estimated Shipment: 05-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16071531
|
enQuireBio™
QP7909-EC-1MG |
1 mg |
26990.00 SEK
1mg |
Estimated Shipment: 05-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16061531
|
enQuireBio™
QP7909-EC-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16081531
|
enQuireBio™
QP7909-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16091531
|
enQuireBio™
QP7909-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16001541
|
enQuireBio™
QP7909-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_001136105.1 | |
PIK3R5 Protein | |
Not determined. | |
Full Length of Isoform 2 | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
23533 | |
Research Use Only | |
PIK3R5 | |
Recombinant Protein | |
E. coli | |
MDSGYVEDSE ESSSEWPWRR GSQERRGHRR PGQKFIRIYK LFKSTSQLVL RRDSRSLEGS SDTALPLRRA GSLCSPLDEP VSPPSRAQRS RSLPQPKLGT QLPSWLLAPA SRPQRRRPFL SGDEDPKAST LRVVVFGSDR ISGKVARAYS NLRRLENNRP LLTRFFKLQF FYVPVKRSHG TSPGACPPPR SQTPSPPTDS PRHASPGELG TTPWEESTND ISHYLGMLDP WYERNVLGLM HLPPEVLCQQ SLKAEAQALE GSPTQLPILA DMLLYYCRFA ARPVLLQVYQ TELTFITGEK TTEIFIHSLE LGHSAATRAI KASGPGSKRL GIDGDREAVP LTLQIIYSKG AISGRSRWSN LEKVCTSVNL NKACRKQEEL DSSMEALTLN LTEVVKRQNS KSKKGFNQIS TSQIKVDKVQ IIGSNSCPFA VCLDQDERKI LQSVVRCEVS PCYKPEKSDL SSPPQTPPDL PAQAAPDLCS LLCLPIMTFS GALP | |
Greater than 90% as determined by SDS-PAGE. |