missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human PLA2G2D / Phospholipase A2 IID Protein
A DNA sequence encoding the Homo sapiens (Human) PLA2G2D / Phospholipase A2 IID, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP8240-ec-10ug
Detalles adicionales : Weight : 0.01000kg
Especificaciones
NP_001258743.1 | |
PLA2G2D / Phospholipase A2 IID Protein | |
Research Use Only | |
PLA2G2D | |
Recombinant Protein | |
E. coli | |
ILNLNKMVKQ VTGKMPILSY WPYGCHCGLG GRGQPKDATD WCCQTHDCCY DHLKTQGCSI YKDYYRYNFS QGNIHCSDKG SWCEQQLCAC DKEVAFCLKR NLDTYQKRLR FYWRPHCRGQ TPGC | |
Greater than 90% as determined by SDS-PAGE. |
26279 | |
10 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |