missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Platelet glycoprotein Ib alpha chain Protein
A DNA sequence encoding the Homo sapiens (Human) Platelet glycoprotein Ib alpha chain, was expressed in the hosts and tags indicated.
1580.00 SEK - 26990.00 SEK
Specifications
Accession Number | NP_000164.5 |
---|---|
Gene ID (Entrez) | 2811 |
Name | Platelet glycoprotein Ib alpha chain Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16010860
|
enQuireBio™
QP6109-EC-100UG |
100 μg |
5980.00 SEK
100µg |
Estimated Shipment: 31-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16030860
|
enQuireBio™
QP6109-EC-1MG |
1 mg |
26990.00 SEK
1mg |
Estimated Shipment: 31-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16020860
|
enQuireBio™
QP6109-EC-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16040860
|
enQuireBio™
QP6109-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16050860
|
enQuireBio™
QP6109-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16060860
|
enQuireBio™
QP6109-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_000164.5 | |
Platelet glycoprotein Ib alpha chain Protein | |
Not determined. | |
Extracellular Domain | |
Human | |
His | |
Tris-based buffer, 50% glycerol |
2811 | |
Research Use Only | |
GP1BA | |
Recombinant Protein | |
E. coli | |
HPICEVSKVA SHLEVNCDKR NLTALPPDLP KDTTILHLSE NLLYTFSLAT LMPYTRLTQL NLDRCELTKL QVDGTLPVLG TLDLSHNQLQ SLPLLGQTLP ALTVLDVSFN RLTSLPLGAL RGLGELQELY LKGNELKTLP PGLLTPTPKL EKLSLANNNL TELPAGLLNG LENLDTLLLQ ENSLYTIPKG FFGSHLLPFA FLHGNPWLCN CEILYFRRWL QDNAENVYVW KQGVDVKAMT SNVASVQCDN SDKFPVYKYP GKGCPTLGDE GDTDLYDYYP EEDTEGDKVR ATRTVVKFPT KAHTTPWGLF YSWSTASLDS QMPSSLHPTQ ESTKEQTTFP PRWTPNFTLH MESITFSKTP KSTTEPTPSP TTSEPVPEPA PNMTTLEPTP SPTTPEPTSE PAPSPTTPEP TPIPTIATSP TILVSATSLI TPKSTFLTTT KPVSLLESTK KTIPELDQPP KLRGVLQGHL ESSRNDPFLH PDFCCLLPL | |
Greater than 90% as determined by SDS-PAGE. |