missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human PLCD4 Protein
A DNA sequence encoding the Homo sapiens (Human) PLCD4, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP8084-ec-10ug
Additional Details : Weight : 0.01000kg
Specifications
NP_116115.1 | |
PLCD4 Protein | |
Research Use Only | |
PLCD4 | |
Recombinant Protein | |
E. coli | |
MASLLQDQLT TDQDLLLMQE GMPMRKVRSK SWKKLRYFRL QNDGMTVWHA RQARGSAKPS FSISDVETIR NGHDSELLRS LAEELPLEQG FTIVFHGRRS NLDLMANSVE EAQIWMRGLQ LLVDLVTSMD HQERLDQWLS DWFQRGDKNQ DGKMSFQEVQ RLLHLMNVEM DQEYAFSLFQ AADTSQSGTL EGEEFVQFYK ALTKRAEVQE LFESFSADGQ KLTLLEFLDF LQEEQKERDC TSELALELID RYEPSDSGKL RHVLSMDGFL SYLCSKDGDI FNPACLPIYQ DMTQPLNHYF ICSSHNTYLV GDQLCGQSSV EGYIRALKRG CRCVEVDVWD GPSGEPVVYH GHTLTSRILF KDVVATVAQY AFQTSDYPVI LSLETHCSWE QQQTMARHLT EILGEQLLST TLDGVLPTQL PSPEELRRKI LVKGKKLTLE EDLEYEEEEA EPELEESELA LESQFETEPE PQEQNLQNKD KKKKSKPILC PALSSLVIYL KSVSFRSFTH SKEHYHFYEI SSFSETKAKR LIKEAGNEFV QHNTWQLSRV YPSGLRTDSS NYNPQELWNA GCQMVAMNMQ TAGLEMDICD GHFRQNGGCG YVLKPDFLRD IQSSFHPEKP ISPFKAQTLL IQVISGQQLP KVDKTKEGSI VDPLVKVQIF GVRLDTARQE TNYVENNGFN PYWGQTLCFR VLVPELAMLR FVVMDYDWKS RNDFIGQYTL PWTCMQQGYR HIHLLSKDGI SLRPASIFVY ICIQEGLEGD ES | |
Greater than 90% as determined by SDS-PAGE. |
84812 | |
10 μg | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |