missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human PLTP Protein
A DNA sequence encoding the Homo sapiens (Human) PLTP, was expressed in the hosts and tags indicated.
2075.00 SEK - 21365.00 SEK
Specifications
Accession Number | NP_001229849.1 |
---|---|
Gene ID (Entrez) | 5360 |
Name | PLTP Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16043790
|
enQuireBio™
QP6507-YE-100UG |
100 μg |
7265.00 SEK
100µg |
Estimated Shipment: 31-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16083790
|
enQuireBio™
QP6507-YE-500UG |
500 μg |
19700.00 SEK
500µg |
Estimated Shipment: 31-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16053790
|
enQuireBio™
QP6507-YE-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16073790
|
enQuireBio™
QP6507-YE-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16093790
|
enQuireBio™
QP6507-YE-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16063790
|
enQuireBio™
QP6507-YE-1MG |
1 mg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_001229849.1 | |
PLTP Protein | |
Not determined. | |
Full Length | |
Human | |
His | |
Tris-based buffer, 50% glycerol |
5360 | |
Research Use Only | |
PLTP | |
Recombinant Protein | |
Yeast | |
EFPGCKIRVT SKALELVKQE GLRFLEQELE TITIPDLRGK EGHFYYNISE VKVTELQLTS SELDFQPQQE LMLQITNASL GLRFRRQLLY WFFYDGGYIN ASAEGVSIRT GLELSRDPAG RMKVSNVSCQ ASVSRMHAAF GGTFKKVYDF LSTFITSGMR FLLNQQICPV LYHAGTVLLN SLLDTVPVRS SVDELVGIDY SLMKDPVAST SNLDMDFRGA FFPLTERNWS LPNRAVEPQL QEEERMVYVA FSEFFFDSAM ESYFRAGALQ LLLVGDKVPH DLDMLLRATY FGSIVLLSPA VIDSPLKLEL RVLAPPRCTI KPSGTTISVT ASVTIALVPP DQPEVQLSSM TMDARLSAKM ALRGKALRTQ LDLRRFRIYS NHSALESLAL IPLQAPLKTM LQIGVMPMLN ERTWRGVQIP LPEGINFVHE VVTNHAGFLT IGADLHFAKG LREVIEKNRP ADVRASTAPT PSTAAV | |
Greater than 90% as determined by SDS-PAGE. |