missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human PPARG Protein
A DNA sequence encoding the Homo sapiens (Human) PPARG, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6518-ye-100ug
Additional Details : Weight : 0.01000kg
Specifications
NP_005028.4 | |
PPARG Protein | |
100 μg | |
Not determined. | |
Full Length of Isoform 1 | |
Human | |
His | |
Tris-based buffer, 50% glycerol |
5468 | |
Greater than 80% as determined by SDS-PAGE. | |
Research Use Only | |
PPARG | |
Recombinant Protein | |
Yeast | |
MGETLGDSPI DPESDSFTDT LSANISQEMT MVDTEMPFWP TNFGISSVDL SVMEDHSHSF DIKPFTTVDF SSISTPHYED IPFTRTDPVV ADYKYDLKLQ EYQSAIKVEP ASPPYYSEKT QLYNKPHEEP SNSLMAIECR VCGDKASGFH YGVHACEGCK GFFRRTIRLK LIYDRCDLNC RIHKKSRNKC QYCRFQKCLA VGMSHNAIRF GRMPQAEKEK LLAEISSDID QLNPESADLR ALAKHLYDSY IKSFPLTKAK ARAILTGKTT DKSPFVIYDM NSLMMGEDKI KFKHITPLQE QSKEVAIRIF QGCQFRSVEA VQEITEYAKS IPGFVNLDLN DQVTLLKYGV HEIIYTMLAS LMNKDGVLIS EGQGFMTREF LKSLRKPFGD FMEPKFEFAV KFNALELDDS DLAIFIAVII LSGDRPGLLN VKPIEDIQDN LLQALELQLK LNHPESSQLF AKLLQKMTDL RQIVTEH |