missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human PPP1CB Protein
A DNA sequence encoding the Homo sapiens (Human) PPP1CB, was expressed in the hosts and tags indicated.
1640.00 SEK - 26990.00 SEK
Specifications
Accession Number | NP_002700.1 |
---|---|
Gene ID (Entrez) | 5500 |
Name | PPP1CB Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16063890
|
enQuireBio™
QP6522-EC-100UG |
100 μg |
5980.00 SEK
100µg |
Estimated Shipment: 07-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16083890
|
enQuireBio™
QP6522-EC-1MG |
1 mg |
26990.00 SEK
1mg |
Estimated Shipment: 07-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16073890
|
enQuireBio™
QP6522-EC-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16093890
|
enQuireBio™
QP6522-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16003900
|
enQuireBio™
QP6522-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16013900
|
enQuireBio™
QP6522-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_002700.1 | |
PPP1CB Protein | |
Not determined. | |
Full Length of Mature Protein | |
Human | |
His | |
Tris-based buffer, 50% glycerol |
5500 | |
Research Use Only | |
PPP1CB | |
Recombinant Protein | |
E. coli | |
ADGELNVDSL ITRLLEVRGC RPGKIVQMTE AEVRGLCIKS REIFLSQPIL LELEAPLKIC GDIHGQYTDL LRLFEYGGFP PEANYLFLGD YVDRGKQSLE TICLLLAYKI KYPENFFLLR GNHECASINR IYGFYDECKR RFNIKLWKTF TDCFNCLPIA AIVDEKIFCC HGGLSPDLQS MEQIRRIMRP TDVPDTGLLC DLLWSDPDKD VQGWGENDRG VSFTFGADVV SKFLNRHDLD LICRAHQVVE DGYEFFAKRQ LVTLFSAPNY CGEFDNAGGM MSVDETLMCS FQILKPSEKK AKYQYGGLNS GRPVTPPRTA NPPKKR | |
Greater than 90% as determined by SDS-PAGE. |