missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human PPP1CB Protein

A DNA sequence encoding the Homo sapiens (Human) PPP1CB, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP6522-ec-100ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16063890

  • 4074.00 SEK / 100µg


Estimated Shipment
12-02-2021
Add to basket

Description

Description

Specifications

Specifications

NP_002700.1
PPP1CB Protein
100 μg
Not determined.
Full Length of Mature Protein
Human
His
Tris-based buffer, 50% glycerol
5500
Greater than 90% as determined by SDS-PAGE.
Research Use Only
PPP1CB
Recombinant Protein
E. coli
ADGELNVDSL ITRLLEVRGC RPGKIVQMTE AEVRGLCIKS REIFLSQPIL LELEAPLKIC GDIHGQYTDL LRLFEYGGFP PEANYLFLGD YVDRGKQSLE TICLLLAYKI KYPENFFLLR GNHECASINR IYGFYDECKR RFNIKLWKTF TDCFNCLPIA AIVDEKIFCC HGGLSPDLQS MEQIRRIMRP TDVPDTGLLC DLLWSDPDKD VQGWGENDRG VSFTFGADVV SKFLNRHDLD LICRAHQVVE DGYEFFAKRQ LVTLFSAPNY CGEFDNAGGM MSVDETLMCS FQILKPSEKK AKYQYGGLNS GRPVTPPRTA NPPKKR
Videos
SDS
Documents