missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human PPP3CC Protein

A DNA sequence encoding the Homo sapiens (Human) PPP3CC, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP6523-ec-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16043900

  • 22920.00 SEK / 1mg
Estimated Shipment: 03-08-2022
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_001230903.1
PPP3CC Protein
1 mg
Not determined.
Partial (see sequence information for more details).
Human
His
Tris-based buffer, 50% glycerol
5533
Greater than 90% as determined by SDS-PAGE.
Research Use Only
PPP3CC
Recombinant Protein
E. coli
SGRRFHLSTT DRVIKAVPFP PTQRLTFKEV FENGKPKVDV LKNHLVKEGR LEEEVALKII NDGAAILRQE KTMIEVDAPI TVCGDIHGQF FDLMKLFEVG GSPSNTRYLF LGDYVDRGYF SIECVLYLWS LKINHPKTLF LLRGNHECRH LTDYFTFKQE CRIKYSEQVY DACMETFDCL PLAALLNQQF LCVHGGMSPE ITSLDDIRKL DRFTEPPAFG PVCDLLWSDP SEDYGNEKTL EHYTHNTVRG CSYFYSYPAV CEFLQNNNLL SIIRAHEAQD AGYRMYRKSQ ATGFPSLITI FSAPNYLDVY NNKAAVLKYE NNVMNIRQFN CSPHPYWLPN FMDVFTWSLP FVGEKVTEML VNVLNICSDD ELISDDEAEG STTVRKEIIR NKIRAIGKMA RVFSILRQES ESVLTLKGLT PTGTLPLGVL SGGKQTIETA TVEAVEAREA IRGFSLQHKI RSFEEARGLD RINERMPPRK DSIHAGGPMK SVTSAHSHAA HRSDQGKKAH S
Videos
SDS
Documents
Certificates
Special Offers

Special Offers