missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human PRKRIR Protein
A DNA sequence encoding the Homo sapiens (Human) PRKRIR, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6530-ec-500ug
Additional Details : Weight : 0.01000kg
Specifications
NP_004696.2 | |
PRKRIR Protein | |
Research Use Only | |
PRKRIR | |
Recombinant Protein | |
E. coli | |
MGQLKFNTSE EHHADMYRSD LPNPDTLSAE LHCWRIKWKH RGKDIELPST IYEALHLPDI KFFPNVYALL KVLCILPVMK VENERYENGR KRLKAYLRNT LTDQRSSNLA LLNINFDIKH DLDLMVDTYI KLYTSKSELP TDNSETVENT | |
Greater than 90% as determined by SDS-PAGE. |
5612 | |
500 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |