missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Programmed cell death protein 1

A DNA sequence encoding the Homo sapiens (Human) Programmed cell death protein 5, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP6475-ec-500ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16063570

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Description

Description

Specifications

Specifications

NP_004699.1
Programmed cell death protein 1
Research Use Only
PDCD5
Recombinant Protein
E. coli
ADEELEALRR QRLAELQAKH GDPGDAAQQE AKHREAEMRN SILAQVLDQS ARARLSNLAL VKPEKTKAVE NYLIQMARYG QLSEKVSEQG LIEILKKVSQ QTEKTTTVKF NRRKVMDSDE DDDY
Greater than 80% as determined by SDS-PAGE.
9141
500 μg
Not determined.
Full Length
Human
GST
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers