missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Programmed cell death protein 1
A DNA sequence encoding the Homo sapiens (Human) Programmed cell death protein 6, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP8484-ec-500ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001254485.1 | |
Programmed cell death protein 1 | |
Research Use Only | |
ALG2 | |
Recombinant Protein | |
E. coli | |
MAAYSYRPGP GAGPGPAAGA ALPDQSFLWN VFQRVDKDRS GVISDTELQQ ALSNGTWTPF NPVTVRSIIS MFDRENKAGV NFSEFTGVWK YITDWQNVFR TYDRDNSGMI DKNELKQALS GFGYRLSDQF HDILIRKFDR QGRGQIAFDD FIQGCIVLQR LTDIFRRYDT DQDGWIQVSY EQYLSMVFSI V | |
Greater than 90% as determined by SDS-PAGE. |
10016 | |
500 μg | |
Not determined. | |
Full Length | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |