missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Programmed cell death protein 1

A DNA sequence encoding the Homo sapiens (Human) Programmed cell death protein 6, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP8484-ec-50ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16094871

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Description

Description

Specifications

Specifications

NP_001254485.1
Programmed cell death protein 1
Research Use Only
ALG2
Recombinant Protein
E. coli
MAAYSYRPGP GAGPGPAAGA ALPDQSFLWN VFQRVDKDRS GVISDTELQQ ALSNGTWTPF NPVTVRSIIS MFDRENKAGV NFSEFTGVWK YITDWQNVFR TYDRDNSGMI DKNELKQALS GFGYRLSDQF HDILIRKFDR QGRGQIAFDD FIQGCIVLQR LTDIFRRYDT DQDGWIQVSY EQYLSMVFSI V
Greater than 90% as determined by SDS-PAGE.
10016
50 μg
Not determined.
Full Length
Human
GST
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers