missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Programmed cell death protein 10 Protein
A DNA sequence encoding the Homo sapiens (Human) Programmed cell death protein 10, was expressed in the hosts and tags indicated.
1640.00 SEK - 26990.00 SEK
Specifications
Accession Number | NP_009148.2 |
---|---|
Gene ID (Entrez) | 11235 |
Name | Programmed cell death protein 10 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16022341
|
enQuireBio™
QP8043-EC-100UG |
100 μg |
5980.00 SEK
100µg |
Estimated Shipment: 05-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16042341
|
enQuireBio™
QP8043-EC-1MG |
1 mg |
26990.00 SEK
1mg |
Estimated Shipment: 05-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16032341
|
enQuireBio™
QP8043-EC-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16052341
|
enQuireBio™
QP8043-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16062341
|
enQuireBio™
QP8043-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16072341
|
enQuireBio™
QP8043-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_009148.2 | |
Programmed cell death protein 10 Protein | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
11235 | |
Research Use Only | |
PDCD10 | |
Recombinant Protein | |
E. coli | |
MRMTMEEMKN EAETTSMVSM PLYAVMYPVF NELERVNLSA AQTLRAAFIK AEKENPGLTQ DIIMKILEKK SVEVNFTESL LRMAADDVEE YMIERPEPEF QDLNEKARAL KQILSKIPDE INDRVRFLQT IKDIASAIKE LLDTVNNVFK KYQYQNRRAL EHQKKEFVKY SKSFSDTLKT YFKDGKAINV FVSANRLIHQ TNLILQTFKT VA | |
Greater than 90% as determined by SDS-PAGE. |