missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Protachykinin-1 Protein
A DNA sequence encoding the Homo sapiens (Human) Protachykinin-1, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6758-ec-10ug
Additional Details : Weight : 0.01000kg
Specifications
NP_003173.1 | |
Protachykinin-1 Protein | |
Research Use Only | |
TAC1 | |
Recombinant Protein | |
E. coli | |
MKILVALAVF FLVSTQLFAE EIGANDDLNY WSDWYDSDQI KEELPEPFEH LLQRIARRPK PQQFFGLMGK RDADSSIEKQ VALLKALYGH GQISHKRHKT DSFVGLMGKR ALNSVAYERS AMQNYERRR | |
Greater than 80% as determined by SDS-PAGE. |
6863 | |
10 μg | |
Not determined. | |
Full Length | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |