missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Protein AATF Protein

A DNA sequence encoding the Homo sapiens (Human) Protein AATF, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP8502-ec-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16044981

  • 26990.00 SEK / 1mg
Estimated Shipment: 31-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_036270.1
Protein AATF Protein
Research Use Only
AATF
Recombinant Protein
E. coli
PLALQLEQLL NPRPSEADPE ADPEEATAAR VIDRFDEGED GEGDFLVVGS IRKLASASLL DTDKRYCGKT TSRKAWNEDH WEQTLPGSSD EEISDEEGSG DEDSEGLGLE EYDEDDLGAA EEQECGDHRE SKKSRSHSAK TPGFSVQSIS DFEKFTKGMD DLGSSEEEED EESGMEEGDD AEDSQGESEE DRAGDRNSED DGVVMTFSSV KVSEEVEKGR AVKNQIALWD QLLEGRIKLQ KALLTTNQLP QPDVFPLFKD KGGPEFSSAL KNSHKALKAL LRSLVGLQE
Greater than 90% as determined by SDS-PAGE.
26574
1 mg
Not determined.
Partial (see sequence information for more details).
Human
GST
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers