missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Protein AATF Protein
A DNA sequence encoding the Homo sapiens (Human) Protein AATF, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP8502-ec-50ug
Additional Details : Weight : 0.01000kg
Specifications
NP_036270.1 | |
Protein AATF Protein | |
Research Use Only | |
AATF | |
Recombinant Protein | |
E. coli | |
PLALQLEQLL NPRPSEADPE ADPEEATAAR VIDRFDEGED GEGDFLVVGS IRKLASASLL DTDKRYCGKT TSRKAWNEDH WEQTLPGSSD EEISDEEGSG DEDSEGLGLE EYDEDDLGAA EEQECGDHRE SKKSRSHSAK TPGFSVQSIS DFEKFTKGMD DLGSSEEEED EESGMEEGDD AEDSQGESEE DRAGDRNSED DGVVMTFSSV KVSEEVEKGR AVKNQIALWD QLLEGRIKLQ KALLTTNQLP QPDVFPLFKD KGGPEFSSAL KNSHKALKAL LRSLVGLQE | |
Greater than 90% as determined by SDS-PAGE. |
26574 | |
50 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |