missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Protein ABHD18 Protein
A DNA sequence encoding the Homo sapiens (Human) Protein ABHD18, was expressed in the hosts and tags indicated.
2515.00 SEK - 38570.00 SEK
Specifications
Name | Protein ABHD18 Protein |
---|---|
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Gene Symbol | C4orf29 |
Protein Length | Full Length |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16059170
|
enQuireBio™
QP7525-EC-100UG |
100 μg |
8695.00 SEK
100µg |
Estimated Shipment: 30-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16079170
|
enQuireBio™
QP7525-EC-1MG |
1 mg |
38570.00 SEK
1mg |
Estimated Shipment: 30-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16069170
|
enQuireBio™
QP7525-EC-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16089170
|
enQuireBio™
QP7525-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16099170
|
enQuireBio™
QP7525-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16009180
|
enQuireBio™
QP7525-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
Protein ABHD18 Protein | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
Research Use Only | |
C4orf29 | |
Recombinant Protein | |
E. coli | |
RPEDLKRLFE FRKMIGNRER CQNLVSSDYP VHIDKIEEQS DCKILDGHFV SPMAHYVPDI MPIESVIARF QFIVPKEWNS KYRPVCIHLA GTGDHHYWRR RTLMARPMIK EARMASLLLE NPYYGCRKPK DQVRSSLKNV SDLFVMGGAL VLESAALLHW LEREGYGPLG MTGISMGGHM ASLAVSNWPK PMPLIPCLSW STASGVFTTT DSFKMGQEFV KHFTSSADKL TNLNLVSRTL NLDISNQVVS QKPADCHNSS KTSVSATSEG LLLQDTSKMK RFNQTLSTNK SGYTSRNPQS YHLLSKEQSR NSLRKESLIF MKGVMDECTH VANFSVPVDP SLIIVVQAKE DAYIPRTGVR SLQEIWPGCE IRYLEGGHIS AYLFKQGLFR | |
Greater than 90% as determined by SDS-PAGE. |