missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Protein-arginine deiminase type-1 Protein
A DNA sequence encoding the Homo sapiens (Human) Protein-arginine deiminase type-1, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP8239-ec-500ug
Additional Details : Weight : 0.01000kg
Specifications
NP_037490.2 | |
Protein-arginine deiminase type-1 Protein | |
500 μg | |
Not determined. | |
Full Length | |
Human | |
His | |
Tris-based buffer, 50% glycerol |
29943 | |
Greater than 90% as determined by SDS-PAGE. | |
Research Use Only | |
PADI1 | |
Recombinant Protein | |
E. coli | |
MAPKRVVQLS LKMPTHAVCV VGVEAHVDIH SDVPKGANSF RVSGSSGVEV FMVYNRTRVK EPIGKARWPL DTDADMVVSV GTASKELKDF KVRVSYFGEQ EDQALGRSVL YLTGVDISLE VDTGRTGKVK RSQGDKKTWR WGPEGYGAIL LVNCDRDNHR SAEPDLTHSW LMSLADLQDM SPMLLSCNGP DKLFDSHKLV LNVPFSDSKR VRVFCARGGN SLSDYKQVLG PQCLSYEVER QPGEQEIKFY VEGLTFPDAD FLGLVSLSVS LVDPGTLPEV TLFTDTVGFR MAPWIMTPNT QPPEELYVCR VMDTHGSNEK FLEDMSYLTL KANCKLTICP QVENRNDRWI QDEMEFGYIE APHKSFPVVF DSPRNRGLKD FPYKRILGPD FGYVTREIPL PGPSSLDSFG NLDVSPPVTV GGTEYPLGRI LIGSSFPKSG GRQMARAVRN FLKAQQVQAP VELYSDWLSV GHVDEFLTFV PTSDQKGFRL LLASPSACLK LFQEKKEEGY GEAAQFDGLK HQAKRSINEM LADRHLQRDN LHAQKCIDWN RNVLKRELGL AESDIVDIPQ LFFLKNFYAE AFFPDMVNMV VLGKYLGIPK PYGPIINGRC CLEEKVQSLL EPLGLHCIFI DDYLSYHELQ GEIHCGTNVR RKPFPFKWWN MVP |