missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Protein atonal homolog 1 Protein

A DNA sequence encoding the Homo sapiens (Human) Protein atonal homolog 1, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP7932-ec-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16011681

  • 26990.00 SEK / 1mg
Estimated Shipment: 24-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_005163.1
Protein atonal homolog 1 Protein
Research Use Only
ATOH1
Recombinant Protein
E. coli
MSRLLHAEEW AEVKELGDHH RQPQPHHLPQ PPPPPQPPAT LQAREHPVYP PELSLLDSTD PRAWLAPTLQ GICTARAAQY LLHSPELGAS EAAAPRDEVD GRGELVRRSS GGASSSKSPG PVKVREQLCK LKGGVVVDEL GCSRQRAPSS KQVNGVQKQR RLAANARERR RMHGLNHAFD QLRNVIPSFN NDKKLSKYET LQMAQIYINA LSELLQTPSG GEQPPPPPAS CKSDHHHLRT AASYEGGAGN ATAAGAQQAS GGSQRPTPPG SCRTRFSAPA SAGGYSVQLD ALHFSTFEDS ALTAMMAQKN LSPSLPGSIL QPVQEENSKT SPRSHRSDGE FSPHSHYSDS DEAS
Greater than 90% as determined by SDS-PAGE.
474
1 mg
Not determined.
Full Length
Human
His-SUMO
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers