missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Protein C8orf4 Protein

A DNA sequence encoding the Homo sapiens (Human) Protein C8orf4, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP8178-ec-50ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16003101

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Description

Description

Specifications

Specifications

NP_064515.1
Protein C8orf4 Protein
Research Use Only
C8orf4
Recombinant Protein
E. coli
MKAKRSHQAV IMSTSLRVSP SIHGYHFDTA SRKKAVGNIF ENTDQESLER LFRNSGDKKA EERAKIIFAI DQDVEEKTRA LMALKKRTKD KLFQFLKLRK YSIKVH
Greater than 80% as determined by SDS-PAGE.
56892
50 μg
Not determined.
Full Length
Human
GST
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers