missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Protein ERGIC-53-like Protein
A DNA sequence encoding the Homo sapiens (Human) Protein ERGIC-53-like, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP8055-ye-1mg
Additional Details : Weight : 0.01000kg
Specifications
NP_068591.2 | |
Protein ERGIC-53-like Protein | |
Research Use Only | |
LMAN1L | |
Recombinant Protein | |
Yeast | |
GCPPLRRFEY KLSFKGPRLA LPGAGIPFWS HHGDAILGLE EVRLTPSMRN RSGAVWSRAS VPFSAWEVEV QMRVTGLGRR GAQGMAVWYT RGRGHVGSVL GGLASWDGIG IFFDSPAEDT QDSPAIRVLA SDGHIPSEQP GDGASQGLGS CHWDFRNRPH PFRARITYWG QRLRMSLNSG LTPSDPGEFC VDVGPLLLVP GGFFGVSAAT GTLADDHDVL SFLTFSLSEP SPEVPPQPFL EMQQLRLARQ LEGLWARLGL GTREDVTPKS DSEAQGEGER LFDLEETLGR HRRILQALRG LSKQLAQAER QWKKQLGPPG QARPDGGWAL DASCQIPSTP GRGGHLSMSL NKDSAKVGAL LHGQWTLLQA LQEMRDAAVR MAAEAQVSYL PVGIEHHFLE LDHILGLLQE ELRGPAKAAA KAPRPPGQPP RASSCLQ | |
Greater than 90% as determined by SDS-PAGE. |
79748 | |
1 mg | |
Not determined. | |
Full Length | |
Human | |
His | |
Tris-based buffer, 50% glycerol |