missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Protein-lysine N-methyltransferase N6AMT2 Protein
A DNA sequence encoding the Homo sapiens (Human) Protein-lysine N-methyltransferase N6AMT2, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP7833-ec-200ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001305868.1 | |
Protein-lysine N-methyltransferase N6AMT2 Protein | |
Research Use Only | |
N6AMT2 | |
Recombinant Protein | |
E. coli | |
SDLEDDETPQ LSAHALAALQ EFYAEQKQQI EPGEDDKYNI GIIEENWQLS QFWYSQETAL QLAQEAIAAV GEGGRIACVS APSVYQKLRE LCRENFSIYI FEYDKRFAMY GEEFIFYDYN NPLDLPERIA AHSFDIVIAD PPYLSEECLR KTSETVKYLT RGKILLCTGA IMEEQAAELL GVKMCTFVPR HTRNLANEFR CYVNYDSGLD CGI | |
Greater than 90% as determined by SDS-PAGE. |
221143 | |
200 μg | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |