missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Protein phosphatase 1 regulatory subunit 15A Protein
A DNA sequence encoding the Homo sapiens (Human) Protein phosphatase 1 regulatory subunit 15A, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP9341-ye-1mg
Additional Details : Weight : 0.01000kg
Specifications
NP_055145.3 | |
Protein phosphatase 1 regulatory subunit 15A Protein | |
Research Use Only | |
PPP1R15A | |
Recombinant Protein | |
Yeast | |
GPLEPWLVEA VKGAALVEAG LEGEARTPLA IPHTPWGRRP EEEAEDSGGP GEDRETLGLK TSSSLPEAWG LLDDDDGMYG EREATSVPRG QGSQFADGQR APLSPSLLIR TLQGSDKNPG EEKAEEEGVA EEEGVNKFSY PPSHRECCPA VEEEDDEEAV KKEAHRTSTS ALSPGSKPST WVSCPGEEEN QATEDKRTER SKGARKTSVS PRSSGSDPRS WEYRSGEASE EKEEKAHKET GKGEAAPGPQ SSAPAQRPQL KSWWCQPSDE EEGEVKALGA AEKDGEAECP PCIPPPSAFL KAWVYWPGED TEEEEDEEED EDSDSGSDEE EGEAEASSST PATGVFLKSW VYQPGEDTEE EEDEDSDTGS AEDEREAETS ASTPPASAFL KAWVYRPGED TEEEEDEDVD SEDKEDDSEA ALGEAESDPH PSHPDQRAHF RGWGYRPGKE TEEEEAAEDW GEAEPCPFRV AIYVPGEKPP PPWAPPRLPL RLQRRLKRPE TPTHDPDPET PLKARKVRFS EKVTVHFLAV WAGPAQAARQ GPWEQLARDR SRFARRITQA QEELSPCLTP AARARAWARL RNPPLAPIPA LTQTLPSSSV PSSPVQTTPL SMS-QAVATP SRSSAAAAAA LDLSGRRG | |
Greater than 90% as determined by SDS-PAGE. |
23645 | |
1 mg | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
His | |
Tris-based buffer, 50% glycerol |