missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Protein SSX4 Protein

A DNA sequence encoding the Homo sapiens (Human) Protein SSX4, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP6733-ec-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16015460

  • 22640.00 SEK / 1mg
Estimated Shipment: 10-11-2023
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_001030004.1
Protein SSX4 Protein
Research Use Only
SSX4
Recombinant Protein
E. coli
MNGDDAFARR PRDDAQISEK LRKAFDDIAK YFSKKEWEKM KSSEKIVYVY MKLNYEVMTK LGFKVTLPPF MRSKRAADFH GNDFGNDRNH RNQVERPQMT FGSLQRIFPK IMPKKPAEEE NGLKEVPEAS GPQNDGKQLC PPGNPSTLEK INKTSGPKRG KHAWTHRLRE RKQLVVYEEI SDPEEDDE
Greater than 80% as determined by SDS-PAGE.
548313
1 mg
Not determined.
Full Length
Human
GST
Tris-based buffer, 50% glycerol
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers