missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Protein SSX4 Protein
A DNA sequence encoding the Homo sapiens (Human) Protein SSX4, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6733-ec-200ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001030004.1 | |
Protein SSX4 Protein | |
Research Use Only | |
SSX4 | |
Recombinant Protein | |
E. coli | |
MNGDDAFARR PRDDAQISEK LRKAFDDIAK YFSKKEWEKM KSSEKIVYVY MKLNYEVMTK LGFKVTLPPF MRSKRAADFH GNDFGNDRNH RNQVERPQMT FGSLQRIFPK IMPKKPAEEE NGLKEVPEAS GPQNDGKQLC PPGNPSTLEK INKTSGPKRG KHAWTHRLRE RKQLVVYEEI SDPEEDDE | |
Greater than 80% as determined by SDS-PAGE. |
548313 | |
200 μg | |
Not determined. | |
Full Length | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |