missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Protein Wnt-3a Protein
A DNA sequence encoding the Homo sapiens (Human) Protein Wnt-3a, was expressed in the hosts and tags indicated.
1580.00 SEK - 18390.00 SEK
Specifications
Accession Number | NP_149122.1 |
---|---|
Gene ID (Entrez) | 89780 |
Name | Protein Wnt-3a Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16046751
|
enQuireBio™
QP8838-EC-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16086751
|
enQuireBio™
QP8838-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16036751
|
enQuireBio™
QP8838-EC-100UG |
100 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16066751
|
enQuireBio™
QP8838-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16076751
|
enQuireBio™
QP8838-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16056751
|
enQuireBio™
QP8838-EC-1MG |
1 mg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_149122.1 | |
Protein Wnt-3a Protein | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
His | |
Tris-based buffer, 50% glycerol |
89780 | |
Research Use Only | |
Wnt3a | |
Recombinant Protein | |
E. coli | |
SYPIWWSLAV GPQYSSLGSQ PILCASIPGL VPKQLRFCRN YVEIMPSVAE GIKIGIQECQ HQFRGRRWNC TTVHDSLAIF GPVLDKATRE SAFVHAIASA GVAFAVTRSC AEGTAAICGC SSRHQGSPGK GWKWGGCSED IEFGGMVSRE FADARENRPD ARSAMNRHNN EAGRQAIASH MHLKCKCHGL SGSCEVKTCW WSQPDFRAIG DFLKDKYDSA SEMVVEKHRE SRGWVETLRP RYTYFKVPTE RDLVYYEASP NFCEPNPETG SFGTRDRTCN VSSHGIDGCD LLCCGRGHNA RAERRREKCR CVFHWCCYVS CQECTRVYDV HTC | |
Greater than 90% as determined by SDS-PAGE. |