missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Protoporphyrinogen oxidase Protein
A DNA sequence encoding the Homo sapiens (Human) Protoporphyrinogen oxidase, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6521-ec-200ug
Additional Details : Weight : 0.01000kg
Spezifikation
NP_000300.1 | |
Protoporphyrinogen oxidase Protein | |
Research Use Only | |
PPOX | |
Recombinant Protein | |
E. coli | |
MGRTVVVLGG GISGLAASYH LSRAPCPPKV VLVESSERLG GWIRSVRGPN GAIFELGPRG IRPAGALGAR TLLLVSELGL DSEVLPVRGD HPAAQNRFLY VGGALHALPT GLRGLLRPSP PFSKPLFWAG LRELTKPRGK EPDETVHSFA QRRLGPEVAS LAMDSLCRGV FAGNSRELSI RSCFPSLFQA EQTHRSILLG LLLGAGRTPQ PDSALIRQAL AERWSQWSLR GGLEMLPQAL ETHLTSRGVS VLRGQPVCGL SLQAEGRWKV SLRDSSLEAD HVISAIPASV LSELLPAEAA PLARALSAIT AVSVAVVNLQ YQGAHLPVQG FGHLVPSSED PGVLGIVYDS VAFPEQDGSP PGLRVTVMLG GSWLQTLEAS GCVLSQELFQ QRAQEAAATQ LGLKEMPSHC LVHLHKNCIP QYTLGHWQKL ESARQFLTAH RLPLTLAGAS YEGVAVNDCI ESGRQAAVSV LGTEPNS | |
Greater than 90% as determined by SDS-PAGE. |
5498 | |
200 μg | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |