missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human PTPN5 Protein
A DNA sequence encoding the Homo sapiens (Human) PTPN5, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6554-ec-500ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001035059.1 | |
PTPN5 Protein | |
Research Use Only | |
PTPN5 | |
Recombinant Protein | |
E. coli | |
LQAEFFEIPM NFVDPKEYDI PGLVRKNRYK TILPNPHSRV CLTSPDPDDP LSSYINANYI RGYGGEEKVY IATQGPIVST VADFWRMVWQ EHTPIIVMIT NIEEMNEKCT EYWPEEQVAY DGVEITVQKV IHTEDYRLRL ISLKSGTEER GLKHYWFTSW PDQKTPDRAP PLLHLVREVE EAAQQEGPHC APIIVHCSAG IGRTGCFIAT SICCQQLRQE GVVDILKTTC QLRQDRGGMI QTCEQYQFVH HVMSLY | |
Greater than 90% as determined by SDS-PAGE. |
84867 | |
500 μg | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |