missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human PTPRR Protein
A DNA sequence encoding the Homo sapiens (Human) PTPRR, was expressed in the hosts and tags indicated.
1640.00 SEK - 26990.00 SEK
Specifications
Name | PTPRR Protein |
---|---|
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Gene Symbol | PTPRR |
Protein Length | Full Length |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16059830
|
enQuireBio™
QP7619-EC-100UG |
100 μg |
5980.00 SEK
100µg |
Estimated Shipment: 31-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16079830
|
enQuireBio™
QP7619-EC-1MG |
1 mg |
26990.00 SEK
1mg |
Estimated Shipment: 31-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16069830
|
enQuireBio™
QP7619-EC-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16089830
|
enQuireBio™
QP7619-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16099830
|
enQuireBio™
QP7619-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16009840
|
enQuireBio™
QP7619-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
PTPRR Protein | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
Research Use Only | |
PTPRR | |
Recombinant Protein | |
E. coli | |
MILHRLKERF QLSLRQDKEK NQEIHLSPIT LQPALSEAKT VHSMVQPEQA PKVLNVVVDP QGRGAPEIKA TTATSVCPSP FKMKPIGLQK RRGSNVSLTL DMSSLGNIEP FVSIPTPREK VAMEYLQSAS RILTRSQLRD VVASSHLLQS EFMEIPMNFV DPKEIDIPRH GTKNRYKTIL PNPLSRVCLR PKNVTDSLST YINANYIRGY SGKEKAFIAT QGPMINTVDD FWQMVWQEDS PVIVMITKLK EKNEKCVLYW PEKRGIYGKV EVLVISVNEC DNYTIRNLVL KQGSHTQHVK HYWYTSWPDH KTPDSAQPLL QLMLDVEEDR LASQGRGPVV VHCSAGIGRT GCFIATSIGC QQLKEEGVVD ALSIVCQLRM DRGGMVQTSE QYEFVHHALC LYESRLSAET VQ | |
Greater than 90% as determined by SDS-PAGE. |