missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Pulmonary surfactant-associated protein 1

A DNA sequence encoding the Homo sapiens (Human) Pulmonary surfactant-associated protein C, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP6682-ye-50ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16015070

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Description

Description

Specifications

Specifications

NP_001165828.1
Pulmonary surfactant-associated protein 1
Research Use Only
SFTPC
Recombinant Protein
Yeast
FGIPCCPVHL KRLLIVVVVV VLIVVVIVGA LLMGL
Greater than 90% as determined by SDS-PAGE.
6440
50 μg
Not determined.
Full Length
Human
His
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers