Learn More
enQuireBio™ Recombinant Human Pulmonary surfactant-associated protein 1
A DNA sequence encoding the Homo sapiens (Human) Pulmonary surfactant-associated protein C, was expressed in the hosts and tags indicated.
Brand:  enQuireBio™ QP6682-ye-1mg
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 16085060
  • 31280.00 SEK / 1mg
Estimated Shipment: 24-06-2024
to see stock.
Specifications
NP_001165828.1
Pulmonary surfactant-associated protein 1
Research Use Only
SFTPC
Recombinant Protein
Yeast
FGIPCCPVHL KRLLIVVVVV VLIVVVIVGA LLMGL
Greater than 90% as determined by SDS-PAGE.
6440
1 mg
Not determined.
Full Length
Human
His
Tris-based buffer, 50% glycerol