missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Pulmonary surfactant-associated protein 1
A DNA sequence encoding the Homo sapiens (Human) Pulmonary surfactant-associated protein C, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6682-ma-100ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001165828.1 | |
Pulmonary surfactant-associated protein 1 | |
100 μg | |
Not determined. | |
Full Length | |
Human | |
His | |
Tris-based buffer, 50% glycerol |
6440 | |
Greater than 90% as determined by SDS-PAGE. | |
Research Use Only | |
SFTPC | |
Recombinant Protein | |
Mammal | |
FGIPCCPVHL KRLLIVVVVV VLIVVVIVGA LLMGL |