missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Pyridoxal phosphate phosphatase Protein
Click to view available options
Quantity:
100 μg
20 μg
50 μg
Unit Size:
100µg
20µg
50µg
Specifications
Specifications
Accession Number | NP_064711.1 |
Gene ID (Entrez) | 57026 |
Name | Pyridoxal phosphate phosphatase Protein |
Quantity | 20 μg |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Gene Symbol | Pdxp |
Protein Length | Extracellular Domain |
Product Type | Recombinant Protein |
Cross Reactivity | Human |
Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
enQuireBio™ Recombinant Human Pyridoxal phosphate phosphatase Protein >
Spot an opportunity for improvement?Share a Content Correction