missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human RAB7A Protein
A cDNA sequence encoding the RAB7A was constructed and used to recombinantly synthesize the protein.
1545.00 SEK - 46290.00 SEK
Specifications
Name | RAB7A Protein |
---|---|
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Product Type | Recombinant Protein |
Cross Reactivity | Human |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15992959
|
enQuireBio™
QP13246-5UG |
5 μg |
1545.00 SEK
5µg |
Estimated Shipment: 14-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15982959
|
enQuireBio™
QP13246-20UG |
20 μg |
2405.00 SEK
20µg |
Estimated Shipment: 20-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15972959
|
enQuireBio™
QP13246-1MG |
1 mg |
46290.00 SEK
1mg |
Estimated Shipment: 20-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
RAB7A Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
The RAB7A protein solution (0.5 mg/ml) is formulated in 20mM Tris-HCl buffer (pH 8.0), 1mM DTT, 100mM NaCl and 30% glycerol. |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MTSRKKVLLK VIILGDSGVG KTSLMNQYVN KKFSNQYKAT IGADFLTKEV MVDDRLVTMQ IWDTAGQERF QSLGVAFYRG ADCCVLVFDV TAPNTFKTLD SWRDEFLIQA SPRDPENFPF VVLGNKIDLE NRQVATKRAQ AWCYSKNNIP YFETSAKEAINVEQAFQTIA RNALKQETEV ELYNEFPEPI KLDKNDRAKA SAESCSC | |
Greater than 90% as determined by SDS-PAGE. |