missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Ragulator complex protein LAMTOR3 Protein
A DNA sequence encoding the Homo sapiens (Human) Ragulator complex protein LAMTOR3, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6343-ec-10ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001230665.1 | |
Ragulator complex protein LAMTOR3 Protein | |
Research Use Only | |
LAMTOR3 | |
Recombinant Protein | |
E. coli | |
MADDLKRFLY KKLPSVEGLH AIVVSDRDGV PVIKVANDNA PEHALRPGFL STFALATDQG SKLGLSKNKS IICYYNTYQV VQFNRLPLVV SFIASSSANT GLIVSLEKEL APLFEELRQV VEVS | |
Greater than 80% as determined by SDS-PAGE. |
8649 | |
10 μg | |
Not determined. | |
Full Length | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |