missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Ras-related protein Rab-3C Protein
A DNA sequence encoding the Homo sapiens (Human) Ras-related protein Rab-3C, was expressed in the hosts and tags indicated.
1580.00 SEK - 26990.00 SEK
Specifications
Accession Number | NP_001304844.1 |
---|---|
Gene ID (Entrez) | 115827 |
Name | Ras-related protein Rab-3C Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16051711
|
enQuireBio™
QP7937-EC-100UG |
100 μg |
5980.00 SEK
100µg |
Estimated Shipment: 31-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16071711
|
enQuireBio™
QP7937-EC-1MG |
1 mg |
26990.00 SEK
1mg |
Estimated Shipment: 31-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16061711
|
enQuireBio™
QP7937-EC-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16081711
|
enQuireBio™
QP7937-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16091711
|
enQuireBio™
QP7937-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16001721
|
enQuireBio™
QP7937-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_001304844.1 | |
Ras-related protein Rab-3C Protein | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
115827 | |
Research Use Only | |
RAB3C | |
Recombinant Protein | |
E. coli | |
MRHEAPMQMA SAQDARYGQK DSSDQNFDYM FKLLIIGNSS VGKTSFLFRY ADDSFTSAFV STVGIDFKVK TVFKNEKRIK LQIWDTAGQE RYRTITTAYY RGAMGFILMY DITNEESFNA VQDWSTQIKT YSWDNAQVIL VGNKCDMEDE RVISTERGQH LGEQLGFEFF ETSAKDNINV KQTFERLVDI ICDKMSESLE TDPAITAAKQ NTRLKETPPP PQPNCAC | |
Greater than 90% as determined by SDS-PAGE. |