missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Ras-related protein Rab-4A Protein
A DNA sequence encoding the Homo sapiens (Human) Ras-related protein Rab-4A, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6570-ec-1mg
Additional Details : Weight : 0.01000kg
Specifications
NP_001258927.1 | |
Ras-related protein Rab-4A Protein | |
1 mg | |
Not determined. | |
Full Length | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |
5867 | |
Greater than 80% as determined by SDS-PAGE. | |
Research Use Only | |
RAB4A | |
Recombinant Protein | |
E. coli | |
MSQTAMSETY DFLFKFLVIG NAGTGKSCLL HQFIEKKFKD DSNHTIGVEF GSKIINVGGK YVKLQIWDTA GQERFRSVTR SYYRGAAGAL LVYDITSRET YNALTNWLTD ARMLASQNIV IILCGNKKDL DADREVTFLE ASRFAQENEL MFLETSALTG ENVEEAFVQC ARKILNKIES GELDPERMGS GIQYGDAALR QLRSPRRAQA PNAQECGC |