missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human RBKS / Ribokinase Protein

A DNA sequence encoding the Homo sapiens (Human) RBKS / Ribokinase, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP6585-ec-500ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16044340

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Description

Description

Specifications

Specifications

NP_001274509.1
RBKS / Ribokinase Protein
Research Use Only
RBKS
Recombinant Protein
E. coli
AASGEPQRQW QEEVAAVVVV GSCMTDLVSL TSRLPKTGET IHGHKFFIGF GGKGANQCVQ AARLGAMTSM VCKVGKDSFG NDYIENLKQN DISTEFTYQT KDAATGTASI IVNNEGQNII VIVAGANLLL NTEDLRAAAN VISRAKVMVC QLEITPATSL EALTMARRSG VKTLFNPAPA IADLDPQFYT LSDVFCCNES EAEILTGLTV GSAADAGEAA LVLLKRGCQV VIITLGAEGC VVLSQTEPEP KHIPTEKVKA VDTTGAGDSF VGALAFYLAY YPNLSLEDML NRSNFIAAVS VQAAGTQSSY PYKKDLPLTL F
Greater than 90% as determined by SDS-PAGE.
64080
500 μg
Not determined.
Full Length
Human
His-SUMO
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers