missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human RELT / TNFRSF19L Protein

A DNA sequence encoding the Homo sapiens (Human) RELT / TNFRSF19L, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP8596-ec-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16015491

  • 22640.00 SEK / 1mg
Estimated Shipment: 06-07-2023
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_116260.2
RELT / TNFRSF19L Protein
1 mg
Not determined.
Partial (see sequence information for more details).
Human
His
Tris-based buffer, 50% glycerol
84957
Greater than 90% as determined by SDS-PAGE.
Research Use Only
RELT
Recombinant Protein
E. coli
STTLWQCPPG EEPDLDPGQG TLCRPCPPGT FSAAWGSSPC QPHARCSLWR RLEAQVGMAT RDTLCGDCWP GWFGPWGVPR VPCQPCSWAP LGTHGCDEWG RRARRGVEVA AGASSGGETR QPGNGTRA
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers