missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Resistin / ADSF / RETN Protein
A DNA sequence encoding the Homo sapiens (Human) Resistin / ADSF / RETN, was expressed in the hosts and tags indicated.
1580.00 SEK - 26990.00 SEK
Specifications
Accession Number | NP_001180303.1 |
---|---|
Gene ID (Entrez) | 56729 |
Name | Resistin / ADSF / RETN Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16044450
|
enQuireBio™
QP6599-EC-100UG |
100 μg |
5980.00 SEK
100µg |
Estimated Shipment: 10-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16064450
|
enQuireBio™
QP6599-EC-1MG |
1 mg |
26990.00 SEK
1mg |
Estimated Shipment: 10-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16054450
|
enQuireBio™
QP6599-EC-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16074450
|
enQuireBio™
QP6599-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16084450
|
enQuireBio™
QP6599-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16094450
|
enQuireBio™
QP6599-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_001180303.1 | |
Resistin / ADSF / RETN Protein | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
56729 | |
Research Use Only | |
RETN | |
Recombinant Protein | |
E. coli | |
KTLCSMEEAI NERIQEVAGS LIFRAISSIG LECQSVTSRG DLATCPRGFA VTGCTCGSAC GSWDVRAETT CHCQCAGMDW TGARCCRVQP | |
Greater than 90% as determined by SDS-PAGE. |