missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Resistin / ADSF / RETN Protein

A DNA sequence encoding the Homo sapiens (Human) Resistin / ADSF / RETN, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP6599-ec-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16064450

  • 18585.00 SEK / 1mg


Estimated Shipment
04-02-2021
Add to basket

Description

Description

Specifications

Specifications

NP_001180303.1
Resistin / ADSF / RETN Protein
1 mg
Not determined.
Full Length
Human
His-SUMO
Tris-based buffer, 50% glycerol
56729
Greater than 90% as determined by SDS-PAGE.
Research Use Only
RETN
Recombinant Protein
E. coli
KTLCSMEEAI NERIQEVAGS LIFRAISSIG LECQSVTSRG DLATCPRGFA VTGCTCGSAC GSWDVRAETT CHCQCAGMDW TGARCCRVQP
Videos
SDS
Documents